2-Chloro-5-methylpyrazine 97%
Fornitore: Apollo Scientific
|
Attenzione
|
OR40049-1G
OR40049-250MG
APOSOR40049-1GEA
172
EUR
APOSOR40049-1G
APOSOR40049-250MG
2-Chloro-5-methylpyrazine 97%
2-Chloro-5-methylpyrazine
Formula:
C₅H₅ClN₂ Peso molecolare: 128,5600 g/mol Temperatura di stoccaggio: Refrigerata |
MDL Number:
MFCD00055039 CAS Number: 59303-10-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...