Stampa…

Ethyl 3-Oxo-3-(2-thienyl)propionate 97%

Fornitore: Apollo Scientific
Attenzione

Synonyms: Ethyl 3-oxo-3-(thiophen-2-yl)propanoate, 3-Oxo-3-(2-thienyl)propionic acid Ethyl ester

OR400136-10G OR400136-5G
APOSOR400136-10GEA 135 EUR
APOSOR400136-10G APOSOR400136-5G
Ethyl 3-Oxo-3-(2-thienyl)propionate 97%
Ethyl 3-Oxo-3-(2-thienyl)propionate
Formula: C₉H₁₀O₃S
Peso molecolare: 198,24 g/mol
Temperatura di stoccaggio: Refrigerata
MDL Number: MFCD00542649
CAS Number: 13669-10-8

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR