2-Isopropoxyphenylboronic acid 95%
Fornitore: Apollo Scientific
Synonyms:
2-Isopropoxybenzeneboronic acid, (2-Isopropoxyphenyl)boronic acid
OR3944-10G
OR3944-1G
APOSOR3944-10GEA
137
EUR
APOSOR3944-10G
APOSOR3944-1G
2-Isopropoxyphenylboronic acid 95%
2-Isopropoxyphenylboronic acid
Formula:
C₉H₁₃BO₃ Peso molecolare: 180,01 g/mol Punto di fusione: 30…35 °C Temperatura di stoccaggio: Refrigerata |
MDL Number:
MFCD03427050 CAS Number: 138008-97-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...