Stampa…

1H-(N-Benzyl)-4-iodopyrazole

Fornitore: Apollo Scientific

Synonyms: 1-Benzyl-4-iodo-1H-pyrazole

OR3757-1G OR3757-5G
APOSOR3757-1GEA 75.7 EUR
APOSOR3757-1G APOSOR3757-5G
1H-(N-Benzyl)-4-iodopyrazole
1H-(N-Benzyl)-4-iodopyrazole
Formula: C₁₀H₉IN₂
Peso molecolare: 284,1 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD04038745
CAS Number: 50877-42-4

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR