Stampa…

2,4-Dibutoxyphenylboronic acid 95+%

Fornitore: Apollo Scientific

OR361100-5G OR361100-1G
APOSOR361100-5GEA 0 EUR
APOSOR361100-5G APOSOR361100-1G
2,4-Dibutoxyphenylboronic acid 95+%
2,4-Dibutoxyphenylboronic acid
Formula: C₁₄H₂₃BO₄
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD07784348
CAS Number: 870778-89-5

Visualizza i codici


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR