1-Decylboronic acid
Fornitore: Apollo Scientific
OR360123-25G
OR360123-5G
APOSOR360123-25GEA
330
EUR
APOSOR360123-25G
APOSOR360123-5G
1-Decylboronic acid
1-Decylboronic acid
Formula:
C₁₀H₂₃BO₂ Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD01074638 CAS Number: 24464-63-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...