2-((Diphenylmethylene)amino)acetonitrile
Fornitore: Apollo Scientific
Synonyms:
N-(Diphenylmethylene)aminoacetonitrile
OR350306-25G
OR350306-5G
OR350306-100G
APOSOR350306-25GEA
81.5
EUR
APOSOR350306-25G
APOSOR350306-5G
APOSOR350306-100G
2-((Diphenylmethylene)amino)acetonitrile
2-((Diphenylmethylene)amino)acetonitrile
Formula:
C₁₅H₁₂N₂ Peso molecolare: 220,2700 g/mol Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD00009970 CAS Number: 70591-20-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...