4-(5-Bromo-2-chlorobenzyl)phenyl ethyl ether
Fornitore: Apollo Scientific
|
Attenzione
|
Synonyms:
4-Bromo-1-chloro-2-(4-ethoxybenzyl)benzene
OR350042-1G
OR350042-5G
APOSOR350042-1GEA
0
EUR
APOSOR350042-1G
APOSOR350042-5G
4-(5-Bromo-2-chlorobenzyl)phenyl ethyl ether
4-(5-Bromo-2-chlorobenzyl)phenyl ethyl ether
Formula:
C₁₅H₁₄BrClO Peso molecolare: 325,64 g/mol Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD11042292 CAS Number: 461432-23-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...