Stampa…

5-Bromo-2-furamide 95+%

Fornitore: Apollo Scientific

Synonyms: 5-Bromofuran-2-carboxamide

OR346696-1G OR346696-5G
APOSOR346696-1GEA 251 EUR
APOSOR346696-1G APOSOR346696-5G
5-Bromo-2-furamide 95+%
5-Bromo-2-furamide
Formula: C₅H₄BrNO₂
Peso molecolare: 190 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00020926
CAS Number: 6134-61-8

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR