Stampa…

2-Bromo-5-phenyl-1,3,4-thiadiazole 95+%

Fornitore: Apollo Scientific

OR346405-10G OR346405-1G OR346405-5G
APOSOR346405-10GEA 528 EUR
APOSOR346405-10G APOSOR346405-1G APOSOR346405-5G
2-Bromo-5-phenyl-1,3,4-thiadiazole 95+%
2-Bromo-5-phenyl-1,3,4-thiadiazole
Formula: C₈H₅BrN₂S
Peso molecolare: 241,11 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD08436697
CAS Number: 53645-95-7

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR