Stampa…

3-(Chloromethyl)-5-(4-methylphenyl)-1,2,4-oxadiazole 95+%

Fornitore: Apollo Scientific

Synonyms: 3-(Chloromethyl)-5-p-tolyl-[1,2,4]oxadiazole

OR346391-1G OR346391-5G
APOSOR346391-1GEA 183 EUR
APOSOR346391-1G APOSOR346391-5G
3-(Chloromethyl)-5-(4-methylphenyl)-1,2,4-oxadiazole 95+%
3-(Chloromethyl)-5-(4-methylphenyl)-1,2,4-oxadiazole
Formula: C₁₀H₉ClN₂O
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD08444979
CAS Number: 73217-33-1

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR