Stampa…

tert-Butyl-3-(chloromethyl)benzoate

Fornitore: Apollo Scientific

Synonyms: 3-Chloromethyl-benzoic acid tert-butyl ester

OR346196-1G OR346196-5G
APOSOR346196-1GEA 62.6 EUR
APOSOR346196-1G APOSOR346196-5G
tert-Butyl-3-(chloromethyl)benzoate
tert-Butyl-3-(chloromethyl)benzoate
Formula: C₁₂H₁₅ClO₂
Peso molecolare: 226,7000 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD02180879
CAS Number: 220510-74-7

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR