Stampa…

2,6-Dichloropyridine-3,4-diamine 98%

Fornitore: Apollo Scientific

Synonyms: 3,4-Diamino-2,6-dichloropyridine

OR345506-1G OR345506-250MG
APOSOR345506-1GEA 149 EUR
APOSOR345506-1G APOSOR345506-250MG
2,6-Dichloropyridine-3,4-diamine 98%
2,6-Dichloropyridine-3,4-diamine
Formula: C₅H₅Cl₂N₃
Peso molecolare: 178,0200 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD10697689
CAS Number: 101079-63-4

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR