Stampa…

2-Acetyl-4-methylpyridine 98%

Fornitore: Apollo Scientific

Synonyms: 1-(4-Methylpyridin-2-yl)ethanone

OR345116-10G OR345116-5G
APOSOR345116-10GEA 105 EUR
APOSOR345116-10G APOSOR345116-5G
2-Acetyl-4-methylpyridine 98%
2-Acetyl-4-methylpyridine
Formula: C₈H₉NO
Peso molecolare: 135,1632 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD01863638
CAS Number: 59576-26-0

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR