Stampa…

2-(4-Bromophenyl)-3-hydroxyacrylonitrile 95%

Fornitore: Apollo Scientific

OR33578-1G OR33578-5G
APOSOR33578-1GEA 270 EUR
APOSOR33578-1G APOSOR33578-5G
2-(4-Bromophenyl)-3-hydroxyacrylonitrile 95%
2-(4-Bromophenyl)-3-hydroxyacrylonitrile
Formula: C₉H₆BrNO
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD12025878
CAS Number: 27956-41-8

Visualizza i codici


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR