3,4-Dihydro-1H-spiro[naphthalene-2,2'-[1,3]thiazolidine]-4'-one 95%
Fornitore: Apollo Scientific
OR33506-1G
OR33506-5G
APOSOR33506-1GEA
234
EUR
APOSOR33506-1G
APOSOR33506-5G
3,4-Dihydro-1H-spiro[naphthalene-2,2'-[1,3]thiazolidine]-4'-one 95%
3,4-Dihydro-1H-spiro[naphthalene-2,2'-[1,3]thiazolidine]-4'-one
Formula:
C₁₂H₁₃NOS Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD16140281 CAS Number: 1221792-37-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...