Stampa…

1-Thia-4-azaspiro[4.4]nonane hydrochloride 95%

Fornitore: Apollo Scientific

OR33499-1G OR33499-5G
APOSOR33499-1GEA 328 EUR
APOSOR33499-1G APOSOR33499-5G
1-Thia-4-azaspiro[4.4]nonane hydrochloride 95%
1-Thia-4-azaspiro[4.4]nonane hydrochloride
Formula: C₇H₁₄ClNS
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD16140324
CAS Number: 1221792-36-4

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR