Stampa…

1-(1,3-Benzoxazol-7-yl)-3-(dimethylamino)-2-propen-1-one 95%

Fornitore: Apollo Scientific

OR33453-1G OR33453-5G
APOSOR33453-1GEA 299 EUR
APOSOR33453-1G APOSOR33453-5G
1-(1,3-Benzoxazol-7-yl)-3-(dimethylamino)-2-propen-1-one 95%
1-(1,3-Benzoxazol-7-yl)-3-(dimethylamino)-2-propen-1-one
Formula: C₁₂H₁₂N₂O₂
Peso molecolare: 216,24 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD09152708
CAS Number: 952182-96-6

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR