5-(1-Phenoxyethyl)-1H-pyrazole 95%
Fornitore: Apollo Scientific
OR33329-1G
OR33329-5G
APOSOR33329-1GEA
343
EUR
APOSOR33329-1G
APOSOR33329-5G
5-(1-Phenoxyethyl)-1H-pyrazole 95%
5-(1-Phenoxyethyl)-1H-pyrazole
Formula:
C₁₁H₁₂N₂O Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD00139123 CAS Number: 321998-96-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...