6-(2-Chloroacetyl)-2H-1,4-benzoxazin-3(4H)-one 95%
Fornitore: Apollo Scientific
Synonyms:
6-(Chloroacetyl)-2H-1,4-benzoxazin-3(4H)-one, 6-(2-Chloroacetyl)-2H-benzo[b][1,4]oxazin-3(4H)-one
OR33306-25G
OR33306-5G
APOSOR33306-25GEA
662
EUR
APOSOR33306-25G
APOSOR33306-5G
6-(2-Chloroacetyl)-2H-1,4-benzoxazin-3(4H)-one 95%
6-(2-Chloroacetyl)-2H-1,4-benzoxazin-3(4H)-one
Formula:
C₁₀H₈ClNO₃ Peso molecolare: 225,63 g/mol Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD01321312 CAS Number: 26518-76-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...