Stampa…

1-(tert-Butyl)-3-methyl-1H-pyrazol-5-ylamine 95%

Fornitore: Apollo Scientific

Synonyms: 1-(tert-Butyl)-3-methyl-1H-pyrazol-5-amine

OR33296-1G OR33296-5G
APOSOR33296-1GEA 82.9 EUR
APOSOR33296-1G APOSOR33296-5G
1-(tert-Butyl)-3-methyl-1H-pyrazol-5-ylamine 95%
1-(tert-Butyl)-3-methyl-1H-pyrazol-5-ylamine
Formula: C₈H₁₅N₃
Peso molecolare: 153,23 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00122731
CAS Number: 141459-53-2

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR