Stampa…

5-Bromo-2-(2,5-dimethyl-1H-pyrrol-1-yl)pyrimidine 97%

Fornitore: Apollo Scientific

OR33273-1G OR33273-5G
APOSOR33273-1GEA 234 EUR
APOSOR33273-1G APOSOR33273-5G
5-Bromo-2-(2,5-dimethyl-1H-pyrrol-1-yl)pyrimidine 97%
5-Bromo-2-(2,5-dimethyl-1H-pyrrol-1-yl)pyrimidine
Formula: C₁₀H₁₀BrN₃
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD02082433
CAS Number: 478258-81-0

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR