1-Benzyl-5-chloro-6-oxo-1,6-dihydro-3-pyridine carboxylic acid 95%
Fornitore: Apollo Scientific
Formula:
C₁₃H₁₀ClNO₃ Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD00232709 CAS Number: 338982-47-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...