4,5-Dibromo-3(2H)-pyridazinone 95%
Fornitore: Apollo Scientific
Synonyms:
4,5-Dibromopyridazin-3(2H)-one
OR33119-1G
OR33119-25G
OR33119-5G
APOSOR33119-1GEA
49.5
EUR
APOSOR33119-1G
APOSOR33119-25G
APOSOR33119-5G
4,5-Dibromo-3(2H)-pyridazinone 95%
4,5-Dibromo-3(2H)-pyridazinone
Formula:
C₄H₂Br₂N₂O Peso molecolare: 253,89 g/mol Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD00023641 CAS Number: 5788-58-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...