Ethyl-4-chloro-8-methoxyquinoline-3-carboxylate 95%
Fornitore: Apollo Scientific
Formula:
C₁₃H₁₂ClNO₃ Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD00173395 CAS Number: 27568-05-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...