Stampa…

Ethyl 2-(2-bromoacetyl)benzenecarboxylate 95%

Fornitore: Apollo Scientific

Synonyms: Ethyl 2-(2-bromoacetyl)benzoate

OR32965-1G OR32965-5G
APOSOR32965-1GEA 538 EUR
APOSOR32965-1G APOSOR32965-5G
Ethyl 2-(2-bromoacetyl)benzenecarboxylate 95%
Ethyl-2-(2-bromoacetyl)benzenecarboxylate
Formula: C₁₁H₁₁BrO₃
Peso molecolare: 271.12 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00142009
CAS Number: 133993-34-7

Visualizza i codici


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR