Stampa…

2-(6,7,8,9-Tetrahydro-5H-[1,2,4]triazolo[4,3-a]azepin-3-yl)benzenol 95%

Fornitore: Apollo Scientific

Synonyms: 2-{5H,6H,7H,8H,9H-[1,2,4]Triazolo[4,3-a]azepin-3-yl}phenol

OR32492-1G OR32492-5G
APOSOR32492-1GEA 234 EUR
APOSOR32492-1G APOSOR32492-5G
2-(6,7,8,9-Tetrahydro-5H-[1,2,4]triazolo[4,3-a]azepin-3-yl)benzenol 95%
2-(6,7,8,9-Tetrahydro-5H-[1,2,4]triazolo[4,3-a]azepin-3-yl)benzenol
Formula: C₁₃H₁₅N₃O
Peso molecolare: 229,28 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD02701039
CAS Number: 108877-44-7

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR