Stampa…

1-(2-Chlorobenzyl)-2-oxo-1,2-dihydro-3-pyridine carboxylic acid 95%

Fornitore: Apollo Scientific

Synonyms: 1-[(2-Chlorophenyl)methyl]-2-oxo-1,2-dihydropyridine-3-carboxylic acid

OR32388-1G OR32388-5G
APOSOR32388-1GEA 234 EUR
APOSOR32388-1G APOSOR32388-5G
1-(2-Chlorobenzyl)-2-oxo-1,2-dihydro-3-pyridine carboxylic acid 95%
1-(2-Chlorobenzyl)-2-oxo-1,2-dihydro-3-pyridine carboxylic acid
Formula: C₁₃H₁₀ClNO₃
Peso molecolare: 263,68 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00140815
CAS Number: 66158-19-8

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR