Methyl-5-methylpyrazole-3-carboxylate 95%
Fornitore: Apollo Scientific
Synonyms:
Methyl 5-methyl-1H-pyrazole-3-carboxylate, Methyl 5-methyl[1H]pyrazole-3-carboxylate, 5-Methylpyrazole-3-carboxylic Acid Methyl Ester
OR32362-1G
OR32362-5G
APOSOR32362-1GEA
58.2
EUR
APOSOR32362-1G
APOSOR32362-5G
Methyl-5-methylpyrazole-3-carboxylate 95%
Methyl-5-methylpyrazole-3-carboxylate
Formula:
C₆H₈N₂O₂ Peso molecolare: 140,14 g/mol Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD03778987 CAS Number: 25016-17-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...