Stampa…

Kartogenin

Fornitore: Apollo Scientific

Synonyms: 2-((4-Phenylphenyl)carbamoyl)benzoic acid, 2-([1,1'-Biphenyl]-4-ylcarbamoyl)benzoic acid, 2-(N-(4-Phenylphenyl)carbamoyl)benzoic acid, N-Biphenyl-4-yl-phthalamic acid

OR323094-250MG OR323094-1G
APOSOR323094-250MGEA 176 EUR
APOSOR323094-250MG APOSOR323094-1G
Kartogenin
Kartogenin
Formula: C₂₀H₁₅NO₃
Peso molecolare: 317,344 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00560510
CAS Number: 4727-31-5

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR