N,N-Dimethyl-2-phenylethanethioamide 95%
Fornitore: Apollo Scientific
Formula:
C₁₀H₁₃NS Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD00051943 CAS Number: 17709-95-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...