1',2',3',4',5',6'-Hexahydro[2,3']bipyridinyl dihydrochloride 95%
Fornitore: Apollo Scientific
|
Attenzione
|
Synonyms:
2-(Piperidin-3-yl)pyridine dihydrochloride
OR321333-1G
OR321333-100MG
OR321333-250MG
APOSOR321333-1GEA
1450
EUR
APOSOR321333-1G
APOSOR321333-100MG
APOSOR321333-250MG
1',2',3',4',5',6'-Hexahydro[2,3']bipyridinyl dihydrochloride 95%
1',2',3',4',5',6'-Hexahydro[2,3']bipyridinyl dihydrochloride
Formula:
C₁₀H₁₆Cl₂N₂ Peso molecolare: 235,16 g/mol Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD14583121 CAS Number: 51747-00-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...