Stampa…

(R)-Benzyl(5-oxotetrahydrofuran-3-yl)carbamate 95+%

Fornitore: Apollo Scientific

Synonyms: (R)-Benzyl (5-oxotetrahydrofuran-3-yl)carbamate, (R)-4-(Benzyloxycarbonylamino)tetrahydrofuran-2-one, (R)-beta-(Cbz-amino)-gamma-butyrolactone, (R)-beta-(Benzyloxycarbonylamino)-gamma-butyrolactone, (R)-beta-(Carbobenzoxyamino)-gamma-butyrolactone, (R)-4-(Carbobenzoxyamino)tetrahydrofuran-2-one, (R)-4-(Cbz-amino)tetrahydrofuran-2-one, Benzyl (R)-Tetrahydro-5-oxo-3-furanylcarbamate

OR318121-1G OR318121-10G OR318121-5G
APOSOR318121-1GEA 61.1 EUR
APOSOR318121-1G APOSOR318121-10G APOSOR318121-5G
(R)-Benzyl(5-oxotetrahydrofuran-3-yl)carbamate 95+%
(R)-Benzyl(5-oxotetrahydrofuran-3-yl)carbamate
Formula: C₁₂H₁₃NO₄
Peso molecolare: 235,2400 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD09839299
CAS Number: 118399-28-3

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR