Stampa…

Methyl-4-(2-aminothiazol-4-yl)benzoate 95+%

Fornitore: Apollo Scientific

Synonyms: Methyl 4-(2-Amino-4-thiazolyl)benzoate

OR318104-1G OR318104-10G OR318104-5G
APOSOR318104-1GEA 288 EUR
APOSOR318104-1G APOSOR318104-10G APOSOR318104-5G
Methyl-4-(2-aminothiazol-4-yl)benzoate 95+%
Methyl-4-(2-aminothiazol-4-yl)benzoate
Formula: C₁₁H₁₀N₂O₂S
Peso molecolare: 234,28 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD03030162
CAS Number: 206555-77-3

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR