Stampa…

2-Cyclopropylbenzylamine 95+%

Fornitore: Apollo Scientific
Pericolo

Synonyms: (2-Cyclopropylphenyl)methanamine

OR318089-1G OR318089-10G OR318089-5G
APOSOR318089-1GEA 488 EUR
APOSOR318089-1G APOSOR318089-10G APOSOR318089-5G
2-Cyclopropylbenzylamine 95+%
2-Cyclopropylbenzylamine
Formula: C₁₀H₁₃N
Peso molecolare: 147,22 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00626840
CAS Number: 118184-66-0
UN: 2735
ADR: 8,III

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR