Stampa…

1-Boc-3-methoxycarbonylmethyleneazetidine 97%

Fornitore: Apollo Scientific

OR317169-1G OR317169-5G
APOSOR317169-1GEA 0 EUR
APOSOR317169-1G APOSOR317169-5G
1-Boc-3-methoxycarbonylmethyleneazetidine 97%
1-Boc-3-methoxycarbonylmethyleneazetidine
Formula: C₁₁H₁₇NO₄
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD13190429
CAS Number: 1105662-87-0

Visualizza i codici


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR