(2S,3aS,7aS)-2-Carboxyoctahydroindole
Fornitore: Apollo Scientific
|
Attenzione
|
Synonyms:
(2S,3aS,7aS)-Octahydro-1H-indole-2-carboxylic acid, H-Oic-OH
OR316048-1G
OR316048-25G
OR316048-5G
APOSOR316048-1GEA
49.5
EUR
APOSOR316048-1G
APOSOR316048-25G
APOSOR316048-5G
(2S,3aS,7aS)-2-Carboxyoctahydroindole
(2S,3aS,7aS)-2-Carboxyoctahydroindole
Formula:
C₉H₁₅NO₂ Peso molecolare: 169,22 g/mol Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD07782125 CAS Number: 80875-98-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...