Stampa…

N-Boc-4-methylene-L-proline

Fornitore: Apollo Scientific

Synonyms: (S)-1-(tert-Butoxycarbonyl)-4-methylenepyrrolidine-2-carboxylic acid

OR316016-1G OR316016-5G
APOSOR316016-1GEA 59.7 EUR
APOSOR316016-1G APOSOR316016-5G
N-Boc-4-methylene-L-proline
N-Boc-4-methylene-L-proline
Formula: C₁₁H₁₇NO₄
Peso molecolare: 227,26 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD01861787
CAS Number: 84348-38-9

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR