N-(tert-Butoxycarbonyl)-L-2-phenylglycinol
Fornitore: Apollo Scientific
Synonyms:
Boc-Phenylglycinol, (S)-tert-Butyl (2-hydroxy-1-phenylethyl)carbamate, N-Boc-L-2-phenylglycinol, (S)-2-(tert-Butoxycarbonylamino)-2-phenylethanol
OR316015-1G
OR316015-25G
OR316015-5G
APOSOR316015-1GEA
49.5
EUR
APOSOR316015-1G
APOSOR316015-25G
APOSOR316015-5G
N-(tert-Butoxycarbonyl)-L-2-phenylglycinol
N-(tert-Butoxycarbonyl)-L-2-phenylglycinol
Formula:
C₁₃H₁₉NO₃ Peso molecolare: 237,30 g/mol Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD00274206 CAS Number: 117049-14-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...