Stampa…

Pimobendan

Fornitore: Apollo Scientific

Synonyms: 6-(2-(4-Methoxyphenyl)-1H-benzo[d]imidazol-5-yl)-5-methyl-4,5-dihydropyridazin-3(2H)-one, 2-(2-Methyl-1-oxopropyl)isoindole-1,3-dione

OR315170-1G OR315170-5G
APOSOR315170-1GEA 77.1 EUR
APOSOR315170-1G APOSOR315170-5G
Pimobendan
Pimobendan
Formula: C₁₉H₁₈N₄O₂
Peso molecolare: 334,38 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00761648
CAS Number: 74150-27-9

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR