2-Amino-3-bromopyrazine 95%
Fornitore: Apollo Scientific
|
Attenzione
|
Formula:
C₄H₄BrN₃ Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD09909692 CAS Number: 21943-12-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...