Stampa…

(2R,3S)-3-Phenylisoserine hydrochloride

Fornitore: Apollo Scientific
Attenzione

OR315015-1G OR315015-5G
APOSOR315015-1GEA 24.4 EUR
APOSOR315015-1G APOSOR315015-5G
(2R,3S)-3-Phenylisoserine hydrochloride
(2R,3S)-3-Phenylisoserine hydrochloride
Formula: C₉H₁₂ClNO₃
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD09750971
CAS Number: 132201-32-2

Visualizza i codici


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR