D-(+)-Malic acid
Fornitore: Apollo Scientific
|
Pericolo
|
Synonyms:
(R)-(+)-Hydroxysuccinic acid, D(+)-Hydroxy-1,4-butanedioic acid, (R)-(+)-Hydroxy-1,4-butanedioic acid, D(+)-Malic acid, D(+)-Hydroxysuccinic acid, (R)-2-Hydroxysuccinic acid
OR315008-100G
OR315008-25G
APOSOR315008-100GEA
84.4
EUR
APOSOR315008-100G
APOSOR315008-25G
D-(+)-Malic acid
D-(+)-Malic acid
Formula:
C₄H₆O₅ Peso molecolare: 134,09 g/mol Punto di fusione: 100…104 °C Densità: 1,6 g/cm³ (25 °C) Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD00004245 CAS Number: 636-61-3 EINECS: 211-262-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...