Stampa…

(R)-(-)-2-Chloromandelic acid

Fornitore: Apollo Scientific

Synonyms: (R)-2-(2-Chlorophenyl)-2-hydroxyacetic acid

OR315005-100G OR315005-25G OR315005-5G
APOSOR315005-100GEA 94.5 EUR
APOSOR315005-100G APOSOR315005-25G APOSOR315005-5G
(R)-(-)-2-Chloromandelic acid
(R)-(-)-2-Chloromandelic acid
Formula: C₈H₇ClO₃
Peso molecolare: 186,59 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00798429
CAS Number: 52950-18-2

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR