Stampa…

tert-Butyl-4-(bromomethyl)benzylcarbamate

Fornitore: Apollo Scientific

Synonyms: tert-Butyl 4-(bromomethyl)benzylcarbamate, N-BOC-4-(Bromomethyl)benzylamine

OR313092-1G OR313092-5G
APOSOR313092-1GEA 94.5 EUR
APOSOR313092-1G APOSOR313092-5G
tert-Butyl-4-(bromomethyl)benzylcarbamate
tert-Butyl-4-(bromomethyl)benzylcarbamate
Formula: C₁₃H₁₈BrNO₂
Peso molecolare: 300,1900 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD11974929
CAS Number: 187283-17-6

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR