Methyl-2-bromo-6-methylisonicotinate
Fornitore: Apollo Scientific
OR313085-25G
OR313085-5G
APOSOR313085-25GEA
303
EUR
APOSOR313085-25G
APOSOR313085-5G
Methyl-2-bromo-6-methylisonicotinate
Methyl-2-bromo-6-methylisonicotinate
Formula:
C₈H₈BrNO₂ Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD04112565 CAS Number: 884494-71-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...