Stampa…

3-(4-Methoxyphenyl)-1H-pyrazole-4-carboxylic acid

Fornitore: Apollo Scientific

OR311037-1G OR311037-500MG
APOSOR311037-1GEA 539 EUR
APOSOR311037-1G APOSOR311037-500MG
3-(4-Methoxyphenyl)-1H-pyrazole-4-carboxylic acid
3-(4-Methoxyphenyl)-1H-pyrazole-4-carboxylic acid
Formula: C₁₁H₁₀N₂O₃
Peso molecolare: 218,21 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD03834503
CAS Number: 618383-46-3

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR