3-Chloro-2,2-bis(chloromethyl)propanoic acid
Fornitore: Apollo Scientific
|
Pericolo
|
OR310319-1G
OR310319-500MG
APOSOR310319-1GEA
270
EUR
APOSOR310319-1G
APOSOR310319-500MG
3-Chloro-2,2-bis(chloromethyl)propanoic acid
3-Chloro-2,2-bis(chloromethyl)propanoic acid
Formula:
C5H7Cl3O2 Peso molecolare: 205,4700 g/mol Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD00021685 CAS Number: 17831-70-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...