Stampa…

2-(Methylthio)-5-pyridineboronic acid pinacol ester

Fornitore: Apollo Scientific

Synonyms: 6-(Methylsulphanyl)pyridine-3-boronic acid pinacol ester, 6-(Methylthio)pyridine-3-boronic acid pinacol ester

OR310224-1G OR310224-500MG
APOSOR310224-1GEA 727 EUR
APOSOR310224-1G APOSOR310224-500MG
2-(Methylthio)-5-pyridineboronic acid pinacol ester
2-(Methylthio)-5-pyridineboronic acid pinacol ester
Formula: C₁₂H₁₈BNO₂S
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD09037495
CAS Number: 849934-89-0

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR