Stampa…

3-Bromoisonicotinonitrile

Fornitore: Apollo Scientific

OR310216-1G OR310216-500MG
APOSOR310216-1GEA 727 EUR
APOSOR310216-1G APOSOR310216-500MG
3-Bromoisonicotinonitrile
3-Bromoisonicotinonitrile
Formula: C₆H₃BrN₂
Peso molecolare: 183,01 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00234143
CAS Number: 13958-98-0

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR